Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCCTGTCAATTGCCTCCTCTTG[A/C]AGGAACATGTGTAGGCAGCGTTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300883 | ||||||||||||||||||||
Literature Links: |
SNX12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX12 - sorting nexin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256185.1 | 580 | Silent Mutation | CTG,CTT | L,L 146 | NP_001243114.1 | |
NM_001256186.1 | 580 | UTR 3 | NP_001243115.1 | |||
NM_001256187.1 | 580 | UTR 3 | NP_001243116.1 | |||
NM_001256188.1 | 580 | Silent Mutation | CTG,CTT | L,L 142 | NP_001243117.1 | |
NM_013346.3 | 580 | Silent Mutation | CTG,CTT | L,L 146 | NP_037478.2 |