Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGCCATTGGGAAGAGGCCTGAC[C/T]GATAGACCATCTCCAAACTGAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300629 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP1S2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP1S2 - adaptor related protein complex 1 sigma 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272071.1 | Intron | NP_001259000.1 | ||||
NM_003916.4 | Intron | NP_003907.3 | ||||
XM_005274614.3 | Intron | XP_005274671.1 | ||||
XM_011545599.1 | Intron | XP_011543901.1 | ||||
XM_017029925.1 | Intron | XP_016885414.1 | ||||
XM_017029926.1 | Intron | XP_016885415.1 |