Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTCGTCTGGGTCAGCACGTAGT[A/T]CATTGGCGAGCATTAATTCTCTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300446 | ||||||||||||||||||||
Literature Links: |
LINC01402 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC01402 - long intergenic non-protein coding RNA 1402 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOXF1 - Rhox homeobox family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139282.2 | 635 | Missense Mutation | GAA,GTA | E,V 170 | NP_644811.1 | |
XM_011531281.2 | 635 | Missense Mutation | GAA,GTA | E,V 198 | XP_011529583.1 |
RHOXF1-AS1 - RHOXF1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |