Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCAGCTGTTCTTGTAGTACCC[A/G]GTATGTATTCCAATATTCTTTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 314997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF75D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF75D - zinc finger protein 75D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185063.2 | 2472 | Intron | NP_001171992.1 | |||
NM_007131.4 | 2472 | Missense Mutation | CGG,TGG | R,W 181 | NP_009062.2 | |
XM_005262469.1 | 2472 | Missense Mutation | CGG,TGG | R,W 181 | XP_005262526.1 | |
XM_005262471.3 | 2472 | UTR 5 | XP_005262528.1 | |||
XM_011531391.1 | 2472 | Missense Mutation | CGG,TGG | R,W 181 | XP_011529693.1 | |
XM_011531395.2 | 2472 | Silent Mutation | ACC,ACT | T,T 61 | XP_011529697.1 | |
XM_011531396.2 | 2472 | Missense Mutation | CGG,TGG | R,W 13 | XP_011529698.1 | |
XM_011531397.2 | 2472 | Silent Mutation | ACC,ACT | T,T 40 | XP_011529699.1 | |
XM_017029818.1 | 2472 | Silent Mutation | ACC,ACT | T,T 65 | XP_016885307.1 | |
XM_017029819.1 | 2472 | Silent Mutation | ACC,ACT | T,T 77 | XP_016885308.1 | |
XM_017029820.1 | 2472 | Silent Mutation | ACC,ACT | T,T 40 | XP_016885309.1 |