Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGTGGTTCCTCTTGCTGAGGCT[C/T]CTCATCACCGGGCTCCTGGAACCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300740 | ||||||||||||||||||||
Literature Links: |
XAGE3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XAGE3 - X antigen family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130776.1 | 217 | Intron | NP_570132.1 | |||
NM_133179.2 | 217 | Missense Mutation | AAG,GAG | K,E 33 | NP_573440.1 | |
XM_011530768.2 | 217 | Missense Mutation | AAG,GAG | K,E 33 | XP_011529070.1 | |
XM_011530769.2 | 217 | Missense Mutation | AAG,GAG | K,E 33 | XP_011529071.1 | |
XM_011530770.2 | 217 | Missense Mutation | AAG,GAG | K,E 33 | XP_011529072.1 |