Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATGATGCCCCTGAAGATGTGCA[C/T]CTAGGGGGCGGCTACCCCGTGGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WDR44 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR44 - WD repeat domain 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184965.1 | 485 | Silent Mutation | CAC,CAT | H,H 18 | NP_001171894.1 | |
NM_001184966.1 | 485 | Silent Mutation | CAC,CAT | H,H 18 | NP_001171895.1 | |
NM_019045.4 | 485 | Silent Mutation | CAC,CAT | H,H 18 | NP_061918.3 | |
XM_011531353.2 | 485 | Silent Mutation | CAC,CAT | H,H 18 | XP_011529655.1 | |
XM_017029600.1 | 485 | UTR 5 | XP_016885089.1 |