Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGAGTTGGTTCATTTTCTGCT[G/C]CTCAAGTATCGAGCCAGGGAGCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300944 MIM: 300177 | ||||||||||||||||||||
Literature Links: |
CSAG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSAG1 - chondrosarcoma associated gene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CSAG4 - CSAG family member 4 (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGEA12 - MAGE family member A12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166386.3 | 717 | Silent Mutation | CTC,CTG | L,L 121 | NP_001159858.1 | |
NM_001166387.3 | 717 | Silent Mutation | CTC,CTG | L,L 121 | NP_001159859.1 | |
NM_005367.6 | 717 | Silent Mutation | CTC,CTG | L,L 121 | NP_005358.2 |