Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCTTGAATGCAGGGAGCCGAAC[A/G]ACTTCCTCGTAGGAAACAACATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300126 MIM: 305360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DKC1 - dyskerin pseudouridine synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPP1 - membrane palmitoylated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166460.1 | 925 | Silent Mutation | GTC,GTT | V,V 257 | NP_001159932.1 | |
NM_001166461.1 | 925 | Silent Mutation | GTC,GTT | V,V 254 | NP_001159933.1 | |
NM_001166462.1 | 925 | Silent Mutation | GTC,GTT | V,V 244 | NP_001159934.1 | |
NM_002436.3 | 925 | Silent Mutation | GTC,GTT | V,V 274 | NP_002427.1 | |
XM_011531167.1 | 925 | Intron | XP_011529469.1 | |||
XM_011531169.1 | 925 | Silent Mutation | GTC,GTT | V,V 148 | XP_011529471.1 |
SNORA56 - small nucleolar RNA, H/ACA box 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |