Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATGTCCATGCTAGGACTGCAGA[A/G]GAACCTCCACAAGAAAAAGCCCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300107 MIM: 300346 | ||||||||||||||||||||
Literature Links: |
BRS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRS3 - bombesin receptor subtype 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HTATSF1 - HIV-1 Tat specific factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163280.1 | 716 | Silent Mutation | GAA,GAG | E,E 98 | NP_001156752.1 | |
NM_014500.4 | 716 | Silent Mutation | GAA,GAG | E,E 98 | NP_055315.2 | |
XM_005262404.3 | 716 | Silent Mutation | GAA,GAG | E,E 98 | XP_005262461.1 |