Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGTGTTTCTCCGGAAGGATCCT[C/T]TTCTGTATACTTGGAGGCTTCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300687 MIM: 300252 | ||||||||||||||||||||
Literature Links: |
ERCC6L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERCC6L - ERCC excision repair 6 like, spindle assembly checkpoint helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017669.2 | 3527 | Missense Mutation | AAG,GAG | K,E 1144 | NP_060139.2 |
PIN4 - peptidylprolyl cis/trans isomerase, NIMA-interacting 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170747.1 | 3527 | Intron | NP_001164218.1 | |||
NM_006223.3 | 3527 | Intron | NP_006214.2 |