Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCTATTTCAGAGGGTCCCCATC[A/G]TTCCTAGAGTACAGGCAGAAATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300284 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB9A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB9A - RAB9A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195328.1 | Intron | NP_001182257.1 | ||||
NM_004251.4 | Intron | NP_004242.1 | ||||
XM_017029964.1 | Intron | XP_016885453.1 |