Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAAGAGTAGAGGATGGGAACTGA[C/T]ACCTAAAGGTGAGACATGGGTCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300627 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF449 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF449 - zinc finger protein 449 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152695.5 | Intron | NP_689908.3 | ||||
XM_011531312.2 | Intron | XP_011529614.1 | ||||
XM_011531313.2 | Intron | XP_011529615.1 | ||||
XM_011531314.2 | Intron | XP_011529616.1 | ||||
XM_017029350.1 | Intron | XP_016884839.1 | ||||
XM_017029351.1 | Intron | XP_016884840.1 | ||||
XM_017029352.1 | Intron | XP_016884841.1 |