Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTGGATTGCGCTGATAGAAAG[A/G]GGTAATTGTACATTTTCAGAAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300439 | ||||||||||||||||||||
Literature Links: |
RNF128 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF128 - ring finger protein 128, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024539.3 | 456 | Silent Mutation | AGA,AGG | R,R 97 | NP_078815.3 | |
NM_194463.1 | 456 | Intron | NP_919445.1 |