Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAGCTCAAAGTGTACAGGACGG[C/T]GACAGAAATTGCAGGCAGCCACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 301300 MIM: 300773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALAS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALAS2 - 5'-aminolevulinate synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000032.4 | 1810 | Missense Mutation | NP_000023.2 | |||
NM_001037967.3 | 1810 | Missense Mutation | NP_001033056.1 | |||
NM_001037968.3 | 1810 | Missense Mutation | NP_001033057.1 | |||
XM_005261995.3 | 1810 | Missense Mutation | XP_005262052.1 | |||
XM_011530771.1 | 1810 | Missense Mutation | XP_011529073.1 | |||
XM_017029354.1 | 1810 | Intron | XP_016884843.1 |
APEX2 - apurinic/apyrimidinic endodeoxyribonuclease 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |