Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACGATGGGCCATTCATGATGAA[C/T]GATGAGGAAGCTTCGGGCGCTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 301870 MIM: 300540 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BGN - biglycan | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001711.5 | 126 | Silent Mutation | AAC,AAT | N,N 37 | NP_001702.1 | |
XM_017029724.1 | 126 | Silent Mutation | AAC,AAT | N,N 37 | XP_016885213.1 |
HAUS7 - HAUS augmin like complex subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |