Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACATGCTCGACCGAGACACTTCC[A/G]GACACACAGGCGGCAGAGCTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300626 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB11 - ankyrin repeat and SOCS box containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012428.2 | 854 | Missense Mutation | CGG,TGG | R,W 275 | NP_001012428.1 | |
NM_001201583.1 | 854 | Missense Mutation | CGG,TGG | R,W 279 | NP_001188512.1 | |
NM_080873.2 | 854 | Missense Mutation | CGG,TGG | R,W 296 | NP_543149.1 |