Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCCGGGATTCCGCCAATTTGGA[A/G]AGCCTGGCCAAACTGGGCATCCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300134 | ||||||||||||||||||||
Literature Links: |
DUSP9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP9 - dual specificity phosphatase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318503.1 | 786 | Silent Mutation | GAA,GAG | E,E 223 | NP_001305432.1 | |
NM_001395.3 | 786 | Silent Mutation | GAA,GAG | E,E 223 | NP_001386.1 | |
XM_011531123.1 | 786 | Silent Mutation | GAA,GAG | E,E 254 | XP_011529425.1 | |
XM_011531124.1 | 786 | Silent Mutation | GAA,GAG | E,E 223 | XP_011529426.1 |