Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCCGGGGGTCTTGGCACCCGG[A/C]GACTGCGAGGTGGTGGTTCCGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRS4 - insulin receptor substrate 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003604.2 | 3899 | Missense Mutation | CGC,CTC | R,L 1220 | NP_003595.1 | |
XM_005262220.2 | 3899 | Missense Mutation | CGC,CTC | R,L 1220 | XP_005262277.1 | |
XM_006724713.3 | 3899 | Missense Mutation | CGC,CTC | R,L 1220 | XP_006724776.1 | |
XM_011531061.1 | 3899 | Missense Mutation | CGC,CTC | R,L 1220 | XP_011529363.1 |
LOC101928358 - uncharacterized LOC101928358 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |