Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACTTGGCAAGTATGAGTTTTACG[C/T]TGACTTAAGTGTTTAGCCATACGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF280C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF280C - zinc finger protein 280C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017666.4 | 2327 | Silent Mutation | CAA,CAG | Q,Q 708 | NP_060136.1 | |
XM_006724765.3 | 2327 | Silent Mutation | CAA,CAG | Q,Q 659 | XP_006724828.1 |