Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGTACAGTTTGCTGAGGAGATC[G/T]TTTGGTGCTTTGCCTTTCTGTTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300208 | ||||||||||||||||||||
Literature Links: |
SCML2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCML2 - sex comb on midleg-like 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006089.2 | 1613 | Silent Mutation | AGA,CGA | R,R 498 | NP_006080.1 | |
XM_006724458.1 | 1613 | Silent Mutation | AGA,CGA | R,R 498 | XP_006724521.1 | |
XM_017029219.1 | 1613 | Silent Mutation | AGA,CGA | R,R 498 | XP_016884708.1 | |
XM_017029220.1 | 1613 | Silent Mutation | AGA,CGA | R,R 498 | XP_016884709.1 | |
XM_017029221.1 | 1613 | Silent Mutation | AGA,CGA | R,R 498 | XP_016884710.1 | |
XM_017029222.1 | 1613 | Silent Mutation | AGA,CGA | R,R 234 | XP_016884711.1 |