Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCAATCCTAATGTGTCCTAGCA[C/T]AGTGATCAAAGCACAGAAAATGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300611 MIM: 300587 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1GALT1C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1GALT1C1 - C1GALT1 specific chaperone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011551.2 | 309 | Missense Mutation | ATG,GTG | M,V 24 | NP_001011551.1 | |
NM_152692.4 | 309 | Missense Mutation | ATG,GTG | M,V 24 | NP_689905.1 |
MCTS1 - MCTS1, re-initiation and release factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |