Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGATTAAAACGGTGTAAAGTCATA[C/T]CAAACCTCAATTTACTTCTCCGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300504 MIM: 300032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATRX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATRX - ATRX, chromatin remodeler | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000489.4 | Intron | NP_000480.3 | ||||
NM_138270.3 | Intron | NP_612114.2 | ||||
XM_005262153.4 | Intron | XP_005262210.2 | ||||
XM_005262154.4 | Intron | XP_005262211.2 | ||||
XM_005262155.4 | Intron | XP_005262212.2 | ||||
XM_005262156.4 | Intron | XP_005262213.2 | ||||
XM_005262157.4 | Intron | XP_005262214.2 | ||||
XM_006724666.3 | Intron | XP_006724729.1 | ||||
XM_006724667.3 | Intron | XP_006724730.1 | ||||
XM_006724668.2 | Intron | XP_006724731.1 | ||||
XM_017029601.1 | Intron | XP_016885090.1 | ||||
XM_017029602.1 | Intron | XP_016885091.1 | ||||
XM_017029603.1 | Intron | XP_016885092.1 | ||||
XM_017029604.1 | Intron | XP_016885093.1 | ||||
XM_017029605.1 | Intron | XP_016885094.1 | ||||
XM_017029606.1 | Intron | XP_016885095.1 | ||||
XM_017029607.1 | Intron | XP_016885096.1 | ||||
XM_017029608.1 | Intron | XP_016885097.1 | ||||
XM_017029609.1 | Intron | XP_016885098.1 | ||||
XM_017029610.1 | Intron | XP_016885099.1 | ||||
XM_017029611.1 | Intron | XP_016885100.1 |