Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCAGAAGATGAAAGCCGAAGA[C/T]ACCAGCAGACGATGGGGAGCTTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 313650 | ||||||||||||||||||||
Literature Links: |
TAF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TAF1 - TATA-box binding protein associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286074.1 | 465 | Intron | NP_001273003.1 | |||
NM_004606.4 | 465 | Intron | NP_004597.2 | |||
NM_138923.3 | 465 | Intron | NP_620278.1 | |||
XM_005262295.1 | 465 | Intron | XP_005262352.1 | |||
XM_005262296.1 | 465 | Intron | XP_005262353.1 | |||
XM_005262297.4 | 465 | Missense Mutation | CAC,TAC | H,Y 124 | XP_005262354.1 | |
XM_005262300.2 | 465 | Intron | XP_005262357.1 | |||
XM_006724682.3 | 465 | Intron | XP_006724745.1 |