Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAGGCCTGTCATGATGATTGAG[A/G]TAGTACATGAGAACGTAGTGAGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300056 MIM: 300552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCCS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCCS - holocytochrome c synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122608.2 | Intron | NP_001116080.1 | ||||
NM_001171991.2 | Intron | NP_001165462.1 | ||||
NM_005333.4 | Intron | NP_005324.3 |
MID1 - midline 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |