Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCCCCTGTGGCTCTCCACAGCT[C/T]GTTCATGGCCATGGTGTAGAAACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300019 MIM: 300013 MIM: 312420 | ||||||||||||||||||||
Literature Links: |
HCFC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCFC1 - host cell factor C1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAA10 - N(alpha)-acetyltransferase 10, NatA catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RENBP - renin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002910.5 | 597 | Missense Mutation | AAG,GAG | K,E 143 | NP_002901.2 | |
XM_017029698.1 | 597 | Missense Mutation | AAG,GAG | K,E 133 | XP_016885187.1 |