Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCTCTCGATCTTGTTCTCAATC[C/T]TGTTCTTAAGGTTGCGCAGGCGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300947 MIM: 300840 MIM: 300526 | ||||||||||||||||||||
Literature Links: |
CCDC120 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC120 - coiled-coil domain containing 120 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRAF2 - PRA1 domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007213.2 | 501 | Missense Mutation | AAG,AGG | K,R 148 | NP_009144.1 |
WDR45 - WD repeat domain 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |