Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTAGCATTAAGCAGTTCCTCTG[A/G]CTGTAGTGAAAACAAGGATGAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300609 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSL3 - male-specific lethal 3 homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193270.2 | 476 | Missense Mutation | GAC,GGC | D,G 122 | NP_001180199.1 | |
NM_001282174.1 | 476 | Intron | NP_001269103.1 | |||
NM_006800.3 | 476 | UTR 5 | NP_006791.2 | |||
NM_078628.1 | 476 | Intron | NP_523352.1 | |||
NM_078629.3 | 476 | Missense Mutation | GAC,GGC | D,G 134 | NP_523353.2 | |
XM_005274440.2 | 476 | UTR 5 | XP_005274497.1 | |||
XM_011545442.2 | 476 | Missense Mutation | GAC,GGC | D,G 134 | XP_011543744.1 |