Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACTTCAGAATGGGCTCATCATA[C/G]TTCTGAATCATAGGCCCGAGCTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300117 | ||||||||||||||||||||
Literature Links: |
FAM133A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM133A - family with sequence similarity 133 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAP1L3 - nucleosome assembly protein 1 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004538.5 | 1372 | Missense Mutation | AAC,AAG | N,K 331 | NP_004529.2 |