Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGCAGCTATGTCATGCACTG[G/T]ATCTGATAAAGGCGACGAGAGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300097 MIM: 300152 MIM: 300153 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAGEB1 - MAGE family member B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGEB3 - MAGE family member B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGEB4 - MAGE family member B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002367.3 | 432 | Missense Mutation | GGA,GTA | G,V 79 | NP_002358.1 |