Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATAATATTAATAAAGGGAGTA[A/G]CTCTTTCAGATCAATGTGGTATTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHROOM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHROOM2 - shroom family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320663.1 | Intron | NP_001307592.1 | ||||
NM_001320664.1 | Intron | NP_001307593.1 | ||||
NM_001649.3 | Intron | NP_001640.1 | ||||
XM_005274500.4 | Intron | XP_005274557.2 | ||||
XM_011545506.1 | Intron | XP_011543808.1 | ||||
XM_017029492.1 | Intron | XP_016884981.1 | ||||
XM_017029493.1 | Intron | XP_016884982.1 | ||||
XM_017029494.1 | Intron | XP_016884983.1 |