Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTCCGCCTCCTGGGTACTGTA[A/G]ATCACCAATTCATGTTGGACTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300970 | ||||||||||||||||||||
Literature Links: |
MORC4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MORC4 - MORC family CW-type zinc finger 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085354.2 | 2723 | Silent Mutation | NP_001078823.1 | |||
NM_024657.4 | 2723 | Silent Mutation | NP_078933.3 | |||
XM_005262190.3 | 2723 | Silent Mutation | XP_005262247.1 | |||
XM_006724691.2 | 2723 | Missense Mutation | XP_006724754.1 | |||
XM_011531027.2 | 2723 | Silent Mutation | XP_011529329.1 | |||
XM_017029844.1 | 2723 | Silent Mutation | XP_016885333.1 |