Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAGCGCCTCGGGCCAGCCTTC[A/G]TACTTGTTGCTGTGCTTACTGTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300492 | ||||||||||||||||||||
Literature Links: |
FAM3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM3A - family with sequence similarity 3 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171132.2 | 1345 | Silent Mutation | TAC,TAT | Y,Y 210 | NP_001164603.1 | |
NM_001171133.2 | 1345 | Silent Mutation | TAC,TAT | Y,Y 172 | NP_001164604.1 | |
NM_001171134.2 | 1345 | Silent Mutation | TAC,TAT | Y,Y 193 | NP_001164605.1 | |
NM_001282311.1 | 1345 | Silent Mutation | TAC,TAT | Y,Y 224 | NP_001269240.1 | |
NM_001282312.1 | 1345 | Silent Mutation | TAC,TAT | Y,Y 210 | NP_001269241.1 | |
NM_021806.3 | 1345 | Silent Mutation | TAC,TAT | Y,Y 210 | NP_068578.2 | |
XM_005274714.3 | 1345 | Silent Mutation | TAC,TAT | Y,Y 231 | XP_005274771.1 | |
XM_005274716.3 | 1345 | Silent Mutation | TAC,TAT | Y,Y 224 | XP_005274773.1 | |
XM_005277879.4 | 1345 | Silent Mutation | TAC,TAT | Y,Y 193 | XP_005277936.1 | |
XM_006724831.2 | 1345 | Silent Mutation | TAC,TAT | Y,Y 232 | XP_006724894.1 | |
XM_006724832.3 | 1345 | Silent Mutation | TAC,TAT | Y,Y 218 | XP_006724895.1 | |
XM_006724833.2 | 1345 | Silent Mutation | TAC,TAT | Y,Y 207 | XP_006724896.1 | |
XM_006724834.2 | 1345 | Silent Mutation | TAC,TAT | Y,Y 177 | XP_006724897.1 | |
XM_011531185.2 | 1345 | Silent Mutation | TAC,TAT | Y,Y 177 | XP_011529487.1 | |
XM_017029699.1 | 1345 | Silent Mutation | TAC,TAT | Y,Y 177 | XP_016885188.1 | |
XM_017029700.1 | 1345 | Silent Mutation | TAC,TAT | Y,Y 169 | XP_016885189.1 | |
XM_017029701.1 | 1345 | Silent Mutation | TAC,TAT | Y,Y 169 | XP_016885190.1 | |
XM_017029702.1 | 1345 | Silent Mutation | TAC,TAT | Y,Y 169 | XP_016885191.1 |