Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTTGGAGCTGTTGGGCGGCAGC[A/G]GCTGCAGCGGCCAGGGACTCAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300897 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC4H2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC4H2 - zinc finger C4H2-type containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178032.2 | 633 | Silent Mutation | GCC,GCT | A,A 136 | NP_001171503.1 | |
NM_001178033.2 | 633 | Intron | NP_001171504.1 | |||
NM_001243804.1 | 633 | Silent Mutation | GCC,GCT | A,A 136 | NP_001230733.1 | |
NM_018684.3 | 633 | Silent Mutation | GCC,GCT | A,A 159 | NP_061154.1 |