Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAATGAACTTGTCACCATCTTC[A/G]ATGTCTTTGCCCTTTGGATTTTTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300304 | ||||||||||||||||||||
Literature Links: |
CUL4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUL4B - cullin 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079872.1 | 2676 | Silent Mutation | ATC,ATT | I,I 784 | NP_001073341.1 | |
NM_003588.3 | 2676 | Silent Mutation | ATC,ATT | I,I 802 | NP_003579.3 | |
XM_005262481.1 | 2676 | Silent Mutation | ATC,ATT | I,I 804 | XP_005262538.1 | |
XM_006724784.1 | 2676 | Silent Mutation | ATC,ATT | I,I 791 | XP_006724847.1 | |
XM_006724785.2 | 2676 | Silent Mutation | ATC,ATT | I,I 789 | XP_006724848.1 | |
XM_011531399.2 | 2676 | Silent Mutation | ATC,ATT | I,I 608 | XP_011529701.1 | |
XM_011531400.2 | 2676 | Silent Mutation | ATC,ATT | I,I 606 | XP_011529702.1 | |
XM_011531401.1 | 2676 | Silent Mutation | ATC,ATT | I,I 590 | XP_011529703.1 |