Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTTGAATAGTTTGGCTACTTCA[C/T]TGAGATCTTCATGCTGCTCACCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300169 MIM: 310490 MIM: 300333 | ||||||||||||||||||||
Literature Links: |
AIFM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AIFM1 - apoptosis inducing factor, mitochondria associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130846.3 | 2024 | Missense Mutation | AAT,AGT | N,S 263 | NP_001124318.2 | |
NM_001130847.3 | 2024 | UTR 3 | NP_001124319.1 | |||
NM_004208.3 | 2024 | Missense Mutation | AAT,AGT | N,S 602 | NP_004199.1 | |
NM_145812.2 | 2024 | Missense Mutation | AAT,AGT | N,S 598 | NP_665811.1 |
RAB33A - RAB33A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004794.2 | 2024 | Intron | NP_004785.1 | |||
XM_017029963.1 | 2024 | Intron | XP_016885452.1 |