Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTAGGCAAGGTCGGCTGCCTG[A/G]TCTGGGGGTGCATCATAGAATTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300959 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXorf36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXorf36 - chromosome X open reading frame 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024689.2 | 662 | Intron | NP_078965.2 | |||
NM_176819.3 | 662 | Silent Mutation | GAC,GAT | D,D 262 | NP_789789.2 | |
XM_005272670.1 | 662 | Silent Mutation | GAC,GAT | D,D 204 | XP_005272727.1 | |
XM_006724559.1 | 662 | Silent Mutation | GAC,GAT | D,D 204 | XP_006724622.1 |