Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTGCCTTTACTAAAGTACCTGA[C/T]GAGGTATGACCCTGCTTGTGGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300314 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF7L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAF7L - TATA-box binding protein associated factor 7 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168474.1 | 803 | Silent Mutation | TCA,TCG | S,S 229 | NP_001161946.1 | |
NM_024885.3 | 803 | Silent Mutation | TCA,TCG | S,S 315 | NP_079161.3 | |
XM_005262152.1 | 803 | Silent Mutation | TCA,TCG | S,S 229 | XP_005262209.1 | |
XM_006724664.1 | 803 | Silent Mutation | TCA,TCG | S,S 315 | XP_006724727.1 |