Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTGTCCATGTCATCCCCCTGG[A/G]ATGTAAGCTCCACACGGGCAGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300688 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCORL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCORL1 - BCL6 corepressor-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184772.2 | Intron | NP_001171701.1 | ||||
NM_021946.4 | Intron | NP_068765.3 | ||||
XM_005262452.4 | Intron | XP_005262509.2 | ||||
XM_005262453.4 | Intron | XP_005262510.2 | ||||
XM_005262454.3 | Intron | XP_005262511.2 | ||||
XM_005262455.4 | Intron | XP_005262512.2 | ||||
XM_005262456.4 | Intron | XP_005262513.2 | ||||
XM_006724776.3 | Intron | XP_006724839.1 | ||||
XM_006724777.3 | Intron | XP_006724840.1 | ||||
XM_006724779.2 | Intron | XP_006724842.1 | ||||
XM_017029721.1 | Intron | XP_016885210.1 | ||||
XM_017029722.1 | Intron | XP_016885211.1 |