Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGGGTGGTGCAAGACCCAAAAC[A/G]GAGGCTCAAGGAATCACAGGGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300921 | ||||||||||||||||||||
Literature Links: |
ARMCX5-GPRASP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMCX5-GPRASP2 - ARMCX5-GPRASP2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199818.1 | 1340 | Silent Mutation | ACA,ACG | T,T 88 | NP_001186747.1 |
BHLHB9 - basic helix-loop-helix domain containing, class B, 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPRASP2 - G protein-coupled receptor associated sorting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004051.3 | 1340 | Silent Mutation | ACA,ACG | T,T 88 | NP_001004051.1 | |
NM_001184874.2 | 1340 | Silent Mutation | ACA,ACG | T,T 88 | NP_001171803.1 | |
NM_001184875.2 | 1340 | Silent Mutation | ACA,ACG | T,T 88 | NP_001171804.1 | |
NM_001184876.2 | 1340 | Silent Mutation | ACA,ACG | T,T 88 | NP_001171805.1 | |
NM_138437.5 | 1340 | Silent Mutation | ACA,ACG | T,T 88 | NP_612446.1 |