Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTCTTCTGTGCCCTGCCAGGGA[C/G]TTGTGGGCTCCTGCGGCCGACACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300197 MIM: 300104 MIM: 300394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6AP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6AP1 - ATPase H+ transporting accessory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001183.5 | 189 | Missense Mutation | GAC,GAG | D,E 55 | NP_001174.2 | |
XM_011531179.1 | 189 | UTR 5 | XP_011529481.1 |
CH17-340M24.3 - uncharacterized protein BC009467 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GDI1 - GDP dissociation inhibitor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAZ - tafazzin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |