Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATCCATGAGTACCAGGGATCTG[C/T]GTATGTCTGCTAATTAGTTGTACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300359 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAGE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAGE1 - sarcoma antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018666.2 | 629 | Missense Mutation | CAT,TAT | H,Y 152 | NP_061136.2 | |
XM_017029621.1 | 629 | Missense Mutation | CAT,TAT | H,Y 152 | XP_016885110.1 | |
XM_017029622.1 | 629 | Intron | XP_016885111.1 | |||
XM_017029623.1 | 629 | Missense Mutation | CAT,TAT | H,Y 152 | XP_016885112.1 | |
XM_017029624.1 | 629 | Missense Mutation | CAT,TAT | H,Y 121 | XP_016885113.1 | |
XM_017029625.1 | 629 | Missense Mutation | CAT,TAT | H,Y 152 | XP_016885114.1 | |
XM_017029626.1 | 629 | Intron | XP_016885115.1 | |||
XM_017029627.1 | 629 | Intron | XP_016885116.1 |