Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTAGGTCTCCGTACAAAGGCATC[A/G]TCTCCGTTCATGGCACCGGGAGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300327 | ||||||||||||||||||||
Literature Links: |
SSX5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SSX5 - SSX family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021015.3 | 100 | Intron | NP_066295.3 | |||
NM_175723.1 | 100 | Intron | NP_783729.1 | |||
XM_011543949.2 | 100 | Silent Mutation | GAC,GAT | D,D 4 | XP_011542251.1 | |
XM_017029758.1 | 100 | Silent Mutation | GAC,GAT | D,D 4 | XP_016885247.1 |