Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCCTAATAGATGATGATATGAA[C/T]GGAGGACAGAAATAGTAATTCATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSIG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VSIG1 - V-set and immunoglobulin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170553.1 | Intron | NP_001164024.1 | ||||
NM_182607.4 | Intron | NP_872413.1 | ||||
XM_005262127.4 | Intron | XP_005262184.1 | ||||
XM_011530936.2 | Intron | XP_011529238.1 |