Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTTCTCTGATGTATAATGAGCTG[C/T]GTTTTCCGAGGGAAGGTCTTCCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300819 | ||||||||||||||||||||
Literature Links: |
ZNF630 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF630 - zinc finger protein 630 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037735.3 | 1811 | Silent Mutation | ACA,ACG | T,T 416 | NP_001032824.2 | |
NM_001190255.2 | 1811 | Silent Mutation | ACA,ACG | T,T 402 | NP_001177184.1 | |
NM_001282201.1 | 1811 | Silent Mutation | ACA,ACG | T,T 416 | NP_001269130.1 | |
NM_001282202.1 | 1811 | Silent Mutation | ACA,ACG | T,T 292 | NP_001269131.1 |
ZNF630-AS1 - ZNF630 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |