Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTTGGGCTGTGTTTACCTGCAA[A/G]CTCCATTTCCTGGGCGGCCATAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300479 | ||||||||||||||||||||
Literature Links: |
FAM9C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM9C - family with sequence similarity 9 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174901.5 | 382 | Missense Mutation | CTT,TTT | L,F 19 | NP_777561.1 | |
XM_005274460.2 | 382 | Intron | XP_005274517.1 | |||
XM_017029321.1 | 382 | Intron | XP_016884810.1 |
LOC105373133 - uncharacterized LOC105373133 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011545614.2 | 382 | Intron | XP_011543916.1 |