Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAACAAAGGAAAAAGTCAGATC[C/T]ACTCAGACCAGAGAGTCAGTGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 301300 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALAS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALAS2 - 5'-aminolevulinate synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000032.4 | 22 | Intron | NP_000023.2 | |||
NM_001037967.3 | 22 | Intron | NP_001033056.1 | |||
NM_001037968.3 | 22 | Intron | NP_001033057.1 | |||
XM_005261995.3 | 22 | Intron | XP_005262052.1 | |||
XM_011530771.1 | 22 | Intron | XP_011529073.1 | |||
XM_017029354.1 | 22 | Intron | XP_016884843.1 |
PAGE2B - PAGE family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015038.2 | 22 | Intron | NP_001015038.1 | |||
XM_011530785.2 | 22 | Intron | XP_011529087.1 | |||
XM_011530786.2 | 22 | UTR 5 | XP_011529088.1 | |||
XM_011530787.2 | 22 | Intron | XP_011529089.1 | |||
XM_017029513.1 | 22 | Intron | XP_016885002.1 |