Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGTGAGCCCTGAGGCAATGAT[C/T]TCAAAATATTTTAAAGAAATATATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300529 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RY10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
P2RY10 - purinergic receptor P2Y10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324218.1 | Intron | NP_001311147.1 | ||||
NM_001324221.1 | Intron | NP_001311150.1 | ||||
NM_001324225.1 | Intron | NP_001311154.1 | ||||
NM_014499.3 | Intron | NP_055314.1 | ||||
NM_198333.2 | Intron | NP_938147.1 |