Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAATGAATCCTGATGAATCACA[A/G]TACTGAGAATTCCCATCTTGTATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300440 MIM: 312180 | ||||||||||||||||||||
Literature Links: |
NKRF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NKRF - NFKB repressing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173487.1 | 1141 | Silent Mutation | TAC,TAT | Y,Y 129 | NP_001166958.1 | |
NM_001173488.1 | 1141 | Silent Mutation | TAC,TAT | Y,Y 114 | NP_001166959.1 | |
NM_017544.3 | 1141 | Silent Mutation | TAC,TAT | Y,Y 114 | NP_060014.2 | |
XM_011531365.2 | 1141 | Silent Mutation | TAC,TAT | Y,Y 114 | XP_011529667.1 |
UBE2A - ubiquitin conjugating enzyme E2 A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |