Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTCCTGGGCCATAAAGTTTCA[C/T]ATCAAAATCACCCCAGAGTTGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300380 MIM: 302020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTPS2 - CTP synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144002.1 | Intron | NP_001137474.1 | ||||
NM_019857.4 | Intron | NP_062831.3 | ||||
NM_175859.2 | Intron | NP_787055.1 | ||||
XM_005274562.3 | Intron | XP_005274619.1 | ||||
XM_006724503.3 | Intron | XP_006724566.1 | ||||
XM_011545545.2 | Intron | XP_011543847.1 |
S100G - S100 calcium binding protein G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004057.2 | Intron | NP_004048.1 | ||||
XM_017029841.1 | Intron | XP_016885330.1 |