Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTGAGGCGCTCGTCGTGGTGGGC[C/T]GACAGGAAGATGAAGTCTGGGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300369 MIM: 300574 | ||||||||||||||||||||
Literature Links: |
ACRC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACRC - acidic repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CXCR3 - C-X-C motif chemokine receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142797.1 | 639 | Silent Mutation | TCA,TCG | S,S 238 | NP_001136269.1 | |
NM_001504.1 | 639 | Silent Mutation | TCA,TCG | S,S 191 | NP_001495.1 | |
XM_005262256.3 | 639 | Silent Mutation | TCA,TCG | S,S 201 | XP_005262313.1 | |
XM_005262257.3 | 639 | Silent Mutation | TCA,TCG | S,S 201 | XP_005262314.1 | |
XM_017029435.1 | 639 | Silent Mutation | TCA,TCG | S,S 238 | XP_016884924.1 | |
XM_017029436.1 | 639 | Silent Mutation | TCA,TCG | S,S 191 | XP_016884925.1 |